Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: MacConkey I Agar

Catalog No. 22349974 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Macconkey I agar; 20/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-349-974 Macconkey I agar; 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-349-974 Supplier BD Supplier No. 215197
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in qualitative procedures for the isolation and cultivation of gram-negative enteric microorganisms from a variety of clinical and nonclinical specimens

  • Ready-to-use
  • Non-sterile
  • Purple highly viscous liquid
  • 100 x 10mm and 100 x 15mm-style plates

Compliance

Meets USP/EP/JP performance specifications.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USP, EP, JP
Color Purple
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type MacConkey I Agar
Product Type Prepared Microbiology Plate
Sterility Non-sterile
Formulation Pancreatic digest of Gelatin 17g/L, Peptones (meat and Casein) 3g/L, Lactose 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Neutral red 0.03g/L, Crystal Violet 0.001g/L, Agar 13.5g/L
For Use With (Application) Gram negative enteric microorganisms
Packaging Type Plate
Quantity 20/Pk.
Weight (Metric) 490 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.