Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: MacConkey II Agar

Catalog No. B21270X Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21270X 100/Pk.
B21172X 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B21270X Supplier BD Supplier No. 221270
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective and differential media for the detection of coliform organisms and enteric pathogens

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Content And Storage 2°C to 8°C
Additive Neutral Red, Crystal Violet, Lactose, Neutral Red Indicator
Detects Coliform Organisms, Enteric pathogens
Form Solid
Manufacturing Origin United States
Media Type MacConkey II Agar
Target Organism Class Gram-negative Bacteria
Test Type Microbial Detection
Formulation Pancreatic digest of Gelatin 17g/L, Pancreatic digest of Casein 1.5g/L, Peptic digest of Animal Tissue 1.5g/L, Lactose 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Neutral red 0.03g/L, Crystal Violet 0.001g/L, Agar 13.5g/L
For Use With (Application) Coliform organisms and enteric pathogens
Incubation Conditions 35 ±2°C, Aerobic
Incubation Time 18 to 24 hr.
Packaging Type Plate
Quantity 100/Pk.
Protein Source Gelatin, Casein, Animal Tissue
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.