Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: MacConkey II Agar with Sorbitol

Catalog No. L97953 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
Media Type:
MacConkey II Agar
MacConkey II Agar with Sorbitol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Media Type
L97953 10/Pk. MacConkey II Agar
L98519 100/Pk. MacConkey II Agar with Sorbitol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. L97953 Supplier BD Supplier No. 297953

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective and differential media for the detection of Escherichia coli serotype O157:H7 associated with hemorrhagic colitis

The media is also used for the isolation of O157:H7 from foods.

  • Ready-to-use
  • Contains chloramphenicol
  • 100 x 10mm and 100 x 15mm-style plates
  • Deep-filled plates reduce effects of drying during prolonged incubation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO, 9000
Color Red
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type MacConkey II Agar
Product Type Prepared Microbiology Plate
Formulation Pancreatic digest of Gelatin 17g/L, Pancreatic digest of Casein 1.5g/L, Peptic digest of Animal Tissue 1.5g/L, D-Sorbitol 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Neutral red 0.03g/L, Crystal Violet 1mg/L
For Use With (Application) Detection of Escherichia coli serotype O157:H7 associated with hemorrhagic colitis, isolation of O157:H7 from foods
Packaging Type Plate
Quantity 10/Pk.
Manufacturing Quality cGMP, FDA-inspected, ISO 9001 Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.