Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Mannitol Salt Agar

Catalog No. B21271X Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
Format:
Plate
Certifications/Compliance:
ISO, 9000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Certifications/Compliance
B21271X 100/Pk. Plate ISO, 9000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B21271X Supplier BD Supplier No. 221271
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the selective isolation and enumeration of staphylococci from clinical and nonclinical materials

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO, 9000
Color Red
Content And Storage 2°C to 8°C
Detects Staphylococci
Form Liquid
Format Plate
Manufacturing Origin United States
Media Type Mannitol Salt Agar
Product Type Prepared Microbiology Plate
Target Organism Class Gram-positive Cocci
Test Type Standard Plate Count for Microorganism
Formulation Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Beef extract 1g/L, Sodium Chloride 75g/L, D-Mannitol 10g/L, Phenol red 25mg/L, Agar 15g/L
For Use With (Application) Staphylococci detection/count
No. of Places 100
Packaging Type Plate
Quantity 100/Pk.
Protein Source Casein, Animal Tissue, Beef
Manufacturing Quality ISO 9000
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.