Learn More
BD BBL™ Prepared Plated Media: Mueller Hinton Agar with 5% Sheep Blood
Shop All BD Diagnostic Systems ProductsDescription
- Ready-to-use
- 100 X 10mm, 100 X 15mm and 150 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Red |
| Country of Origin | United States |
| Form | Solid |
| Format | Plate |
| Manufacturing Origin | United States |
| Media Type | Mueller Hinton Agar with 5% Sheep Blood |
| Product Type | Prepared Microbiology Plate |
| Formulation | Beef extract 2g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L, Agar 17g/L, Sheep Blood, defibrinated 5% |
| For Use With (Application) | Antimicrobial disc diffusion susceptibility testing of Streptococcus pneumoniae with selected agents |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.