Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Mueller Hinton II Agar

Catalog No. B21800X Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
24/Pk.
Packaging Type:
100 x 15mm Plate
150 x 15mm Plate
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
B21800X 24/Pk. 150 x 15mm Plate
B21275X 100/Pk. 100 x 15mm Plate
B21177X 20/Pk. 100 x 15mm Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. B21800X Supplier BD Supplier No. 221800
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For antimicrobial disc diffusion susceptibility testing of common, rapidly growing bacteria by the Bauer-Kirby method

  • Ready-to-use
  • 100 x 10mm, 100 x 15mm and 150 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Beige
Content And Storage 2°C to 8°C
Additive Thymine, Thymidine, Calcium, Magnesium
Form Solid
Manufacturing Origin United States
Media Type Mueller Hinton II Agar
Product Type Prepared Microbiology Plate
Formulation Beef extract 2g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L, Agar 17g/L
For Use With (Application) Antimicrobial disc diffusion susceptibility testing of common rapidly growing bacteria by the Bauer-Kirby method
Packaging Type 150 x 15mm Plate
Quantity 24/Pk.
Dimensions (Dia. x H) 150 x 15 mm
Protein Source Beef, Casein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.