Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Phenylethyl Alcohol Agar with 5% Sheep Blood

Catalog No. B21277X Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21277X 100/Pk.
B21179X 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B21277X Supplier BD Supplier No. 221277
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the isolation of gram-positive organisms, particularly gram-positive cocci, from specimens of mixed gram-positive and gram-negative flora and for determination of hemolytic reactions.

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Red
Content And Storage 2°C to 8°C
Format Mono-Plate Format
Manufacturing Origin United States
Media Type Phenylethyl Alcohol Agar with 5% Sheep Blood
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, beta-Phenylethyl Alcohol 2.5g/L, Agar 15g/L, Sheep Blood, defibrinated 5%
For Use With (Application) Gram positive organisms, particularly gram positive cocci, from mixed specimens
Packaging Type Plate
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.