Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Pseudosel™ Agar (Cetrimide Agar)

Catalog No. b97882x Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
B97882X ISO/DIS, 11014
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B97882X Supplier BD Supplier No. 297882
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the selective isolation and presumptive identification of Pseudomonas aeruginosa from clinical and nonclinical specimens

  • Ready-to-use
  • 100 X 10mm and 100 X 15mm style plates

Compliance

Meets USP/EP/JP performance specifications.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Cream
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Pseudosel agar (Cetrimide agar)
Formulation Pancreatic digest of Gelatin 20g/L, Potassium Sulfate 10g/L, Magnesium Chloride 1.4g/L, Agar 13.6g/L, Cetrimide 0.3g/L, Glycerol 10mL
For Use With (Application) Selective isolation and presumptive identification of Pseudomonas aeruginosa from clinical and nonclinical specimens
Packaging Type Plate
Quantity 10/Pk.
Weight (Metric) 810 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.