Learn More
Description
- Ready-to-use
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Light Yellow |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | R2A Agar |
| Product Type | Prepared Microbiology Plate |
| Target Organism Class | Fastidious Organisms |
| Test Type | Standard Plate Count for Microorganism |
| Formulation | Yeast Extract - 0.5 g, Proteose Peptone No. 3 - 0.5 g, Casamino Acids - 0.5 g, Dextrose - 0.5 g, Soluble Starch - 0.5 g, Sodium Pyruvate - 0.3 g, Dipotassium Phosphate - 0.3 g, Magnesium Sulfate - 0.05 g, Agar - 15.0 g |
| For Use With (Application) | Isolation of Microorganisms |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.