Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Sabouraud Brain Heart Infusion Agar (Deep Fill)

Catalog No. L97802 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
Media Type:
Sabouraud Brain heart infusion Agar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Media Type
L97802 10/Pk. Sabouraud Brain heart infusion Agar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L97802 Supplier BD Supplier No. 297802
This item is not returnable. View return policy

Used in qualitative procedures for cultivation of dermatophytes and other pathogenic and nonpathogenic fungi from clinical and nonclinical specimens

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates
  • Deep-filled plates reduce effects of drying during prolonged incubation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Sabouraud Brain heart infusion Agar
Formulation Brain heart infusion from (solids) 4g/L, Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 10.5g/L, Dextrose 21g/L, Sodium Chloride 2.5g/L, Disodium Phosphate 1.25g/L, Agar 15g/L
For Use With (Application) Cultivation of dermatophytes and other pathogenic and nonpathogenic fungi
Packaging Type Plate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.