Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Sabouraud Dextrose Agar, Emmons (Deep Fill)

Catalog No. L21867 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L21867 100/Pk.
L4321849 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. L21867 Supplier BD Supplier No. 221867
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For cultivation of dermatophytes and other pathogenic and nonpathogenic fungi

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Content And Storage 2°C to 8°C
Detects Fungi, Dermatophytes
Form Solid
Format Plate
Manufacturing Origin United States
Media Type Sabouraud Dextrose Agar, Emmons
Product Type Prepared Microbiology Plate
Target Organism Class Fungi
Test Type Microbial Cultivation
Formulation Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 20g/L, Agar 17g/L
For Use With (Application) Cultivation of dermatophytes and other pathogenic and nonpathogenic fungi
Packaging Type Plate
Quantity 100/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.