Learn More
BD BBL™ Prepared Plated Media: Sabouraud Dextrose Agar, Emmons (Deep Fill)
Shop All BD Diagnostic Systems ProductsDescription
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Light Beige |
| Content And Storage | 2°C to 8°C |
| Form | Solid |
| Manufacturing Origin | United States |
| Media Type | Sabouraud Dextrose Agar, Emmons |
| Formulation | Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 20g/L, Agar 17g/L |
| For Use With (Application) | Cultivation of dermatophytes and other pathogenic and nonpathogenic fungi |
| Packaging Type | Plate |
| Quantity | 20/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.