Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Salmonella Shigella Agar

Catalog No. B21181X Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
20/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21181X 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B21181X Supplier BD Supplier No. 221181
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Differentially selective media for the isolation of pathogenic enteric bacilli, especially those belonging to the genus Salmonella

This media is not recommended for the primary isolation of Shigella.

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Form Liquid
Manufacturing Origin United States
Media Type Salmonella Shigella agar
Formulation Beef extract 5g/L, Pancreatic digest of Casein 2.5g/L, Peptic digest of Animal Tissue 2.5g/L, Lactose 10g/L, Bile Salts 8.5g/L, Sodium Citrate 8.5g/L, Sodium Thiosulfate 8.5g/L, Ferric Citrate 1g/L, Neutral red 0.025g/L, Agar 13.5g/L, Brilliant green 0.330mg/L
For Use With (Application) Enteric bacilli, especially those belonging to the genus Salmonella
Packaging Type Plate
Quantity 20/Pk.
Weight (Metric) 810 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.