Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Seven H11 Agar (Deep Fill)

Catalog No. L21870 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
L21870 ISO/DIS, 11014
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L21870 Supplier BD Supplier No. 221870
This item is not returnable. View return policy

Used in qualitative procedures for the isolation and cultivation of mycobacteria

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates
  • Deep-filled plates reduce the effects of drying during prolonged incubation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Gray
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Seven H11 agar
Formulation Pancreatic digest of Casein 1g/L, Mag/Lnesium Sulfate 0.05g/L, Ferric Ammonium Citrate 0.04g/L, Sodium Citrate 0.4g/L, Ammonium Sulfate 0.5g/L, Monosodiumg/Llutamate 0.5g/L, Disodium Phosphate 1.5g/L, Monopotassium Phosphate 1.5g/L, Agar 13.5g/L, Sodium Chloride 0.85g/L, Dextrose 2g/L, Bovine Albumin V 5g/L, Catalase 3g/L, Pyridoxine 1g/L, Zinc Sulfate 1g/L, Copper Sulfate 1g/L, Biotin 0.5g/L, Calcium Chloride 0.5g/L, Malachite green 0.25g/L, Oleic Acid 0.06g/L, Glycerol 5mL
For Use With (Application) Mycobacteria
Packaging Type Plate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.