Learn More
BD BBL™ Prepared Plated Media: Sorbitol MacConkey Agar with Cefixime and Tellurite
Shop All BD Diagnostic Systems ProductsDescription
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Red |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Sorbitol MacConkey agar with Cefixime and Tellurite |
| Formulation | Pancreatic digest of Gelatin 17g/L, Pancreatic digest of Casein 1.5g/L, Peptic digest of Animal Tissue 1.5g/L, D-Sorbitol 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Neutral red 0.03g/L, Crystal Violet 0.001g/L, Agar 13.5g/L, Cefixime 0.05g/L, Potassium Tellurite 2.5mg/L |
| For Use With (Application) | Detection of Escherichia coli serotype O157:H7 |
| Packaging Type | Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.