Learn More
Description
The media is inhibited the growth of viridans streptococci, other beta-hemolytic and nonhemolytic streptococci, most Enterobacteriaceae, Neisseria species and some Pseudomonas species.
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Light Red |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | SXT Blood Agar |
| Formulation | Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, growth Factors 1.5g/L, Sulfamethoxazole 23.75mg/L, Trimethoprim 1.25mg/L, Sheep Blood, defibrinated 5% |
| For Use With (Application) | Lancefield groups A and B streptococci from throat cultures and other specimens |
| Packaging Type | Plate |
| Quantity | 20/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.