Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: SXT Blood Agar

Catalog No. p-4766795 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
20/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L21809 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L21809 Supplier BD Supplier No. 221809

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

For isolation of Lancefield groups A and B streptococci from throat cultures and other specimens

The media is inhibited the growth of viridans streptococci, other beta-hemolytic and nonhemolytic streptococci, most Enterobacteriaceae, Neisseria species and some Pseudomonas species.

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Red
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type SXT Blood Agar
Formulation Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, growth Factors 1.5g/L, Sulfamethoxazole 23.75mg/L, Trimethoprim 1.25mg/L, Sheep Blood, defibrinated 5%
For Use With (Application) Lancefield groups A and B streptococci from throat cultures and other specimens
Packaging Type Plate
Quantity 20/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.