Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Thayer Martin, Modified (MTM II) Agar

Catalog No. B21568X Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21568X 100/Pk.
B21567X 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21568X Supplier BD Supplier No. 221568

missing translation for 'validMainframeMsg'

For the isolation of pathogenic Neisseria from specimens containing mixed flora of bacteria and fungi

  • Ready for immediate use

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Dark Brown
Content And Storage 2°C to 8°C
Country of Origin United States
Additive Vancomycin, Colistin, Nystatin, Trimethoprim Lactate, IsoVitaleX Enrichment, Vitamin B12, L-Glutamine, Adenine, Guanine Hydrochloride, p-Aminobenzoic Acid, Nicotinamide Adenine Dinucleotide, Thiamine Pyrophosphate, Ferric Nitrate, Thiamine Hydrochloride, L-Cysteine Hydrochloride, L-Cystine, Dextrose
Detects Pathogenic Neisseria, Bacteria, Fungi
Form Liquid
Format Prepared Plated Media
Manufacturing Origin United States
Media Type Thayer Martin, modified (MTM II) Agar
Product Type Prepared Microbiology Plate
Sterility Sterile
Target Organism Class Gram-negative Bacteria
Test Type Microbial Detection
For Use With (Application) Isolation of pathogenic Neisseria, Cultivation of Neisseria species
Incubation Conditions 35 ±2 °C, Aerobic
Incubation Time 18 to 24 hr.
Packaging Type Plate
Quantity 100/Pk.
Protein Source Casein, Meat
Manufacturing Quality CLSI Standards
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.