Learn More
Description
- Ready-to-use
- Enriched with Columbia agar base supplemented with 5% human blood
- 100 x 10mm and 100 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Light Beige |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | V Agar (for G. vaginalis) |
| Formulation | Pancreatic digest of Casein 12g/L, Peptic digest of Animal Tissue 5g/L, Beef extract 3g/L, Yeast extract 3g/L, Peptone 10g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Human Blood, anticoagulated 5% |
| For Use With (Application) | Gardnerella vaginalis from clinical specimens |
| Packaging Type | Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.