Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Violet Red Bile Glucose Agar

Catalog No. 22349975 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Violet red bile glucose agar; 20/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-349-975 Violet red bile glucose agar; 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-349-975 Supplier BD Supplier No. 215198
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used to test for bile-tolerant Gram-negative bacteria

  • Ready-to-use
  • Highly viscous liquid
  • Nonsterile
  • Violet red
  • 100 x 10mm and 100 x 15mm-style plates
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Red
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Violet red bile glucose agar
Sterility Non-sterile
Target Organism Class Gram-negative Bacteria
Test Type Microbial Detection
Incubation Time 18 to 24 hr.
Packaging Type Plate
Quantity 20/Pk.
Manufacturing Quality ISO 9001:2008 Registered, FDA-registered, ISO 13485:2003 Registered
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.