Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: XLD Agar

Catalog No. B4321284X Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4321284X 100/Pk.
B21192X 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B4321284X Supplier BD Supplier No. 221284
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective and differential media for isolation and differentiation of enteric pathogens, especially Shigella and Salmonella species

  • Ready-to-use
  • Inhibitory to gram-positive organisms
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Orange
Content And Storage 2°C to 8°C
Additive Sodium Desoxycholate, Sodium Thiosulfate (H2S indicator), Ferric Ammonium Citrate
Detects Enteric Pathogens, Shigella species, Salmonella species
Form Liquid
Manufacturing Origin United States
Media Type XLD Agar
Target Organism Class Gram-negative Microorganisms
Test Type Microbial Detection
Formulation Xylose 3.5g/L, L-Lysine 5g/L, Lactose 7.5g/L, Saccharose 7.5g/L, Sodium Chloride 5g/L, Yeast extract 3g/L, Phenol red 0.08g/L, Sodium Desoxycholate 2.5g/L, Sodium Thiosulfate 6.8g/L, Ferric Ammonium Citrate 0.8g/L, Agar 13.5g/L
For Use With (Application) Enteric pathogens, especially Shigella species
Incubation Conditions 35 ±2°C, Aerobic
Incubation Time 18 to 24 hr.
Packaging Type Plate
Quantity 100/Pk.
Protein Source Yeast Extract
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.