Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared RODAC™ and Contact Plate for Critical Environments

Catalog No. b21288x Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Trypticase Soy Agar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
B21288X Trypticase Soy Agar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B21288X Supplier BD Supplier No. 221288
This item is not returnable. View return policy

Recommended for Aerobic Plate Count (Microbial Test Limit) for water-miscible cosmetics containing preservatives

  • Trypticase™ Soy Agar with Lecithin and Polysorbate 80 (RODAC™).
      Designed for surface sampling for microbial contamination
    • Contains Trypticase™ Soy Agar with Lecithin and Polysorbate 80
    • Marked grids facilitate counting
    • Inward sloping walls and convex bottom keep agar bed in place during transit and use

Order Info

20 per sleeve

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Beige
Form Solid
Manufacturing Origin United States
Media Type Trypticase™ Soy Agar with Lecithin and Polysorbat
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Lecithin 0.7g/L, Polysorbate 80 5g/L, Agar 15g/L
For Use With (Application) Detection and enumeration of microorganisms present on surfaces of sanitary importance
Packaging Type Plate
Quantity 20/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.