Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Tubed Media-Dubos Medium Albumin

Catalog No. B215334 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10 x 20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B215334 10 x 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B215334 Supplier BD Supplier No. 215334
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used with Dubos Broth Base for cultivating pure cultures of Mycobacterium tuberculosis

  • Tubes contain 5% solution of albumin fraction V from bovine plasma and 7.5% dextrose in normal saline

TRUSTED_SUSTAINABILITY

Specifications

Media Type Dubos Medium Albumin
Formulation 5% soln. of albumin fraction V from bovine plasma and 7.5% dextrose in normal saline
For Use With (Application) Used with Dubos Broth Base for cultivating pure cultures of Mycobacterium tuberculosis
Quantity 10 x 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.