Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prompt™ Inoculation System

Catalog No. B26306 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
60 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26306 60 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B26306 Supplier BD Supplier No. 226306

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Includes

60 Saline tubes and 60 precision wands (for 60 tests)

For the rapid, inoculum standardization of 0.5 McFarland turbidity level of bacterial inocula using Kirby-Bauer procedures for antimicrobial susceptibility testing

  • Requires no incubation, no turbidity adjustments, no dilutions
  • Standardized bacterial suspension can be prepared in less than 60 seconds and held up to six hours at room temperature before use
  • Procedure has a proven high correlation with conventional Kirby-Bauer techniques

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 27°C
Form Solid
Product Type Inoculation system
For Use With (Application) Used for prepare a McFarland turbidity level of bacterial inocula using Bauer-Kirby procedures for antimicrobial susceptibility testing
Includes 60 Saline tubes, 60 wands
Quantity 60 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.