Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Sabouraud Dextrose Agar with Lecithin and Polysorbate 80 Sterile Pack Media - RODAC™ Plates

Catalog No. L21233 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
SDA, w/Lecithin and Polysorbate 80, Rodac Plate; 10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
L21233 SDA, w/Lecithin and Polysorbate 80, Rodac Plate; 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L21233 Supplier BD Supplier No. 221233
This item is not returnable. View return policy

Plated media in gamma-irradiated packs for isolator systems—multiwrap packaging protects media from vaporized hydrogen peroxide exposure

Rodac (Replicate Organism Detection and Counting) plates for surface sampling.

To use:

  • Place wrapped package of plates in the isolator before running the vaporized hydrogen peroxide decontamination cycle
  • When the isolator aeration cycle has finished, remove the two outermost wrappings, then the sterile inner wrapping
  • Use the included inner bag to transport plates from your critical environment

Compliance

Tested for performance and sterility according to Association for the Advancement of Medical Instrumentation (AAMI) guidelines.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO, 9000
Color Light Beige
Content And Storage 2°C to 8°C
Format Rodac
Manufacturing Origin United States
Media Type Sabouraud Dextrose Agar with Lecithin and Polysorb 80
Sterility Sterile
Formulation Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Lecithin 0.7g/L, Polysorbate 80 5g/L, Agar 15g/L
For Use With (Application) Detection and enumeration of microorganisms present on surfaces of sanitary importance
Packaging Type Plate
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.