Learn More
BD BBL™ Sabouraud Dextrose Agar Sterile Pack Media - Settling Plates
Catalog No. 22349906 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Sabouraud Dextrose agar; 100/Pk.
Catalog No.
22-349-906 Sabouraud Dextrose agar; 100/Pk.
1 options
Catalog No. 22-349-906 Supplier BD Supplier No. 292336
Add to Cart
Add to Cart
Gamma-irradiated plated media individually wrapped, then double wrapped in Tyvek™/ polyethylene bags
  • For critical environments
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO/DIS, 11014
Color White
Form Liquid
Manufacturing Origin United States
Media Type Sabouraud Dextrose agar
Sterility Sterile
Packaging Type Plate
Quantity 100/Pk.