Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cefazolin12
GSA_VA
Catalog No. B31593 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Concentration:
30 μg
Catalog No. Concentration
B31593 30 μg
1 options
This item is not returnable. View return policy
Catalog No. B31593 Supplier BD Supplier No. 231593
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Cefazolin12
Concentration 30 μg
Packaging Type Disc
Quantity 10 Pack
For in vitro diagnostic use