Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cefixime1414
Catalog No. L31664 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Concentration:
5 μg
Catalog No. Concentration
L31664 5 μg
1 options
This item is not returnable. View return policy
Catalog No. L31664 Supplier BD Supplier No. 231664
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Cefixime14
Concentration 5 μg
Packaging Type Disc
Quantity 10 Pack
For in vitro diagnostic use