Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Ertapenem86
Catalog No. B32175 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
1 Pack
10 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32175 10 Pack
B32174 1 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B32175 Supplier BD Supplier No. 232175
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Ertapenem 86
Concentration 10 μg
Packaging Type Disc
Quantity 10 Pack
For in vitro diagnostic use