Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Levofloxacin42
Catalog No. B4331705 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Concentration:
5 μg
Catalog No. Concentration
B4331705 5 μg
1 options
This item is not returnable. View return policy
Catalog No. B4331705 Supplier BD Supplier No. 231705
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Levofloxacin
Concentration 5 μg
Packaging Type Disc
Quantity Each
For in vitro diagnostic use