Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Streptomycin
Catalog No. B4331694 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Concentration:
300 μg
Catalog No. Concentration
B4331694 300 μg
1 options
This item is not returnable. View return policy
Catalog No. B4331694 Supplier BD Supplier No. 231694
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Streptomycin
Concentration 300 μg
Packaging Type Disc
Quantity 1 Pack
For in vitro diagnostic use