Learn More
BD BBL™ Sensi-Disc™ Elution Discs for Susceptibility Testing of Anaerobes
Catalog No. B31605 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Concentration:
80 μg
Catalog No. Concentration
B31605 80 μg
1 options
This item is not returnable. View return policy
Catalog No. B31605 Supplier BD Supplier No. 231605
Add to Cart
Add to Cart
This item is not returnable. View return policy
For determining the in vitro susceptibility of anaerobic bacteria to metronidazole by the broth-disc method
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Metronidazole 78
Concentration 80 μg
Packaging Type Disc
Quantity 10 Pack
For in vitro Diagnostic Use.