Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Sensi-Disc™ Single-Disc Dispenser

Catalog No. L60457 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L60457 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L60457 Supplier BD Supplier No. 260457
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Replaces the manual placement of discs using sterile forceps

  • Made of durable 24 gauge stainless steel

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
For Use With (Equipment) Sensi-Disc™ antibiotic susceptibility test disc
Material Stainless Steel
Packaging Type Disc
Quantity Each

For in vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.