Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Disc Amikacin2, AN-30
GSA_VA
Catalog No. B31597 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Concentration:
30 μg
Catalog No. Concentration
B31597 30 μg
1 options
This item is not returnable. View return policy
Catalog No. B31597 Supplier BD Supplier No. 231597
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cost-effective method uses antimicrobial agents impregnated on high-quality paper
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
  • Employs Bauer-Kirby disc diffusion testing method
  • Use in self-tamping dispensers
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
  • Cartridges of 50 discs each are sold individually or in packs of 10
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Amikacin
Concentration 30 μg
Packaging Type Disc
Quantity 10/Pk.