Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Disc Bacitracin, B-2
Catalog No. B31267 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Concentration:
2U
Catalog No. Concentration
B31267 2U
1 options
This item is not returnable. View return policy
Catalog No. B31267 Supplier BD Supplier No. 231267
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cost-effective method uses antimicrobial agents impregnated on high-quality paper
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
  • Employs Bauer-Kirby disc diffusion testing method
  • Use in self-tamping dispensers
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
  • Cartridges of 50 discs each are sold individually or in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Bacitracin
Concentration 2U
Packaging Type Disc
Quantity 10 Cartridges