Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Gentamicin
Catalog No. B31227 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
1 Cartridge (50 discs)
10/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31227 1 Cartridge (50 discs)
B31299 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B31227 Supplier BD Supplier No. 231227
 Restricted Item
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Gentamicin
Concentration 10 μg
Packaging Type Disc
Quantity 1 Cartridge (50 discs)