Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Moxifloxacin83
Catalog No. 14959803 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Concentration:
5 μg
Catalog No. Concentration
14-959-803 5 μg
1 options
This item is not returnable. View return policy
Catalog No. 14-959-803 Supplier BD Supplier No. 231758
Add to Cart
Add to Cart
This item is not returnable. View return policy
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Moxifloxacin83
Concentration 5 μg
Packaging Type Disc
Quantity 10 Pack