Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Neomycin (30μg)
Catalog No. B31313 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Concentration:
30 μg
Catalog No. Concentration
B31313 30 μg
1 options
This item is not returnable. View return policy
Catalog No. B31313 Supplier BD Supplier No. 231313
Add to Cart
Add to Cart
This item is not returnable. View return policy
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Neomycin
Concentration 30 μg
Packaging Type Disc
Quantity 10 Pack