Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Nitrofurantoin (100μg)
Catalog No. B31292 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Concentration:
100 μg
Catalog No. Concentration
B31292 100 μg
1 options
This item is not returnable. View return policy
Catalog No. B31292 Supplier BD Supplier No. 231292
Add to Cart
Add to Cart
This item is not returnable. View return policy
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Nitrofurantoin
Concentration 100 μg
Packaging Type Disc
Quantity 10 Pack