Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Oxytetracycline
Catalog No. B31342 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Concentration:
30 μg
Catalog No. Concentration
B31342 30 μg
1 options
This item is not returnable. View return policy
Catalog No. B31342 Supplier BD Supplier No. 231342
 Restricted Item
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Oxytetracycline
Concentration 30 μg
Packaging Type Disc
Quantity 10 Pack