Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Piperacillin
Catalog No. B31609 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
1/Pk.
10/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31609 10/Pk.
B31608 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B31609 Supplier BD Supplier No. 231609
Add to Cart
Add to Cart
This item is not returnable. View return policy
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Clearly marked on both sides with letters and numbers designating agent and concentration
  • Available in pack of 1 and 10
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Piperacillin
Concentration 100 μg
Packaging Type Disc
Quantity 10/Pk.