Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Sulfamethoxazole-Trimethoprim
Catalog No. B31536 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Concentration:
23.75 μg/1.25 μg
23.75/1.25 μg
Quantity:
1/Pk.
10/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration Quantity
B31536 23.75/1.25 μg 1/Pk.
B31539 23.75 μg/1.25 μg 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B31536 Supplier BD Supplier No. 231536
Add to Cart
Add to Cart
This item is not returnable. View return policy
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Sulfamethoxazole, Trimethoprim
Concentration 23.75/1.25 μg
Packaging Type Disc
Quantity 1/Pk.