Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Ticarcillin
Catalog No. B31619 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Concentration:
75 μg
Catalog No. Concentration
B31619 75 μg
1 options
This item is not returnable. View return policy
Catalog No. B31619 Supplier BD Supplier No. 231619
 Restricted Item
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Ticarcillin
Concentration 75 μg
Packaging Type Disc
Quantity 10/Pk.