Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Vancomycin (30mcg)
Catalog No. B31034 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
1/Pk.
10 Cartridges
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31034 1/Pk.
B31353 10 Cartridges
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B31034 Supplier BD Supplier No. 231034
Add to Cart
Add to Cart
This item is not returnable. View return policy
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Vancomycin
Concentration 30 μg
Packaging Type Disc
Quantity 1/Pk.