Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ SEPTI-CHEK™ Blood Culture System: Bottles

Catalog No. L4343189 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
70 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L4343189 70 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L4343189 Supplier BD Supplier No. 243189

missing translation for 'validMainframeMsg'

Designed for rapid qualitative detection, isolation, and differentiation of pathogens from blood

  • Closed, biphasic system
  • Bottles contain one of four specific growth media containing 0.05% sodium polyanethole-sulfonate (SPS) in a CO2 atmosphere, and feature two options in fill volumes (for adult or pediatric applications)
  • Risk of contamination and sample mix-up is minimized

TRUSTED_SUSTAINABILITY

Specifications

Format Bottle
Product Type Microbiology Media
For Use With (Equipment) Septi-Check
Quantity 70 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.