Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Taxo™ Differentiation Discs, Kanamycin
GSA_VA

Catalog No. B31562 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Concentration:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
B31562 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B31562 Supplier BD Supplier No. 231562
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

For use in the presumptive identification of gram-negative anaerobic bacilli

  • 0.25 in. absorbent paper impregnated with accurately determined amounts of antimicrobial agents
  • Last disc in each cartridge is marked X and contains the drug as coded

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Kanamycin
Concentration 1 mg
Content And Storage -20°C to +8°C
Packaging Type Disc
Quantity 50 Discs

For in vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.