Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Taxo™ Differentiation ONPG Discs

Catalog No. B31248 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Concentration:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
B31248 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B31248 Supplier BD Supplier No. 231248
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For detection of lactose fermenters

  • Useful for those that do not promptly ferment lactose in some routine identification media such as Triple Sugar Iron Agar (TSI Agar) or Kligler Iron Agar
  • 6mm discs made from high-quality absorbent pape
  • Impregnated with approximately 100μg of o-nitrophenyl-β-D-galactopyranoside per disc

Order Info

Vial of 50 discs.

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents o-nitrophenyl-beta-D-galactopyranoside
Concentration 100 μg
Content And Storage -20°C to +8°C
For Use With (Application) Detecting lactose fermenters
Packaging Type Vial
Quantity 50 Discs

For in vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.