Learn More
BD BBL™ Taxo™ X Factor Differentaition Discs
Catalog No. B231729 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
1 Pack
Catalog No. Quantity
B231729 1 Pack
1 options
This item is not returnable. View return policy
Catalog No. B231729 Supplier BD Supplier No. 231729
Add to Cart
Add to Cart
This item is not returnable. View return policy
For differentiating Haemophilus species
  • 0.25 in. diameter
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents X Factor
Content And Storage -20°C to +8°C
Packaging Type Disc
Quantity 1 Pack