Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Trypticase™ Soy Agar w/5% Sheep Blood/Levine EMB Agar, I Plate™ Prepared Media

Catalog No. L21289 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L21289 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L21289 Supplier BD Supplier No. 221289
Add to Cart
Add to Cart
This item is not returnable. View return policy

Two media formulations in single plate for cultivating fastidious microorganisms

Tripticase Soy Agar (TSA II)

  • For the visualization of hemolytic reactions produced by many bacterial species
Levine EMB Agar
  • Selective and differential plating medium for isolation of gram-negative enteric bacteria

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Dark Yellow
Form Liquid
Manufacturing Origin United States
Media Type Trypticase™ Soy Agar with 5% Sheep Blood (TSA II)/Levine EMB Agar
For Use With (Application) Cultivation of fastidious microorganisms and for visualization of hemolytic reaction
Packaging Type Plate
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.