Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Trypticase™ Soy Agar w/5% Sheep Blood/ MacConkey II, I Plate™ Prepared Media

Catalog No. B21291X Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
Media Type:
Trypticase™ Soy Agar (TSA II), MacConkey II Agar
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Media Type
B21291X 100/Pk. Trypticase™ Soy Agar (TSA II), MacConkey II Agar
B21290X 20/Pk. Trypticase™ Soy Agar (TSA II), MacConkey II Agar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B21291X Supplier BD Supplier No. 221291
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Two media formulations in single plate for cultivating fastidious microorganisms

Tripticase Soy Agar (TSA II)

  • For the visualization of hemolytic reactions produced by many bacterial species
MacConkey II Agar
  • Selective and differential plating medium for the detection of coliform organisms and enteric pathogens

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2°C to 8°C
Additive Defibrinated Sheep Blood, Bile Salts, Neutral Red, Crystal Violet
Detects Fastidious Microorganisms, Hemolytic Reactions, Coliform Organisms, Enteric Pathogens
Form Liquid
Manufacturing Origin United States
Media Type Trypticase™ Soy Agar (TSA II), MacConkey II Agar
Target Organism Class Fastidious Organisms
Test Type Microbial Cultivation
Formulation Trypticase™ Soy Agar with 5% Sheep Blood (TSA II): Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L, Defibrinated Sheep Blood 5% ; MacConkey II Agar: Pancreatic Digest of Gelatin 17.0g/L, Sodium Chloride 5.0g/L, Pancreatic Digest of Casein 1.5g/L, Neutral Red 0.03g/L, Peptic Digest of Animal Tissue 1.5g/L,Crystal Violet 0.001g/L, Lactose 10.0g/L, Agar13.5g/L, Bile Salts 1.5g/L
For Use With (Application) Cultivation of fastidious microorganisms and visualization of hemolytic reactions
Incubation Time 18 to 24 hr.
No. of Places 2
Packaging Type Plate
Quantity 100/Pk.
Protein Source Pancreatic Digest of Casein, Papaic Digest of Soybean Meal, Defibrinated Sheep Blood, Pancreatic Digest of Gelatin, Peptic Digest of Animal Tissue
Source Sheep
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.